Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Animal-free IFN-lambda 2/IL-28A protein, human (His) | >95.0% | 10UG
SDP

Catalog No. 5000178172
Encompass_Preferred

Recombinant human interferon lambda-2 (IL-28A) with a C-terminal 6xHis tag, expressed in E. coli and supplied as a lyophilized powder. The preparation is endotoxin-low and biologically active for in vitro antiviral and immunomodulatory assays, intended for research and cell culture applications.

  • Species: human.
  • Expression system: E. coli.
  • Tag: C-terminal 6xHis.
  • Formulation: lyophilized from PBS (pH 8.0) with trehalose.
  • Purity: >95% by reducing SDS-PAGE.
  • Predicted molecular mass: 20.6 kDa (≈20 kDa by SDS-PAGE).
  • Endotoxin: <0.1 EU per μg.
  • Biological activity: ED50 <20 ng/mL in protection assay.
  • Recommended use: for research and cell culture only.
  • Storage: store at -20°C; reconstituted material stable at 4°C for 1 week.

Catalog No. 50-001-78172 Supplier Medchemexpress LLC Supplier No. HYP700119AF10UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human interferon lambda-2 (IL-28A) with a C-terminal 6xHis tag, expressed in E. coli and supplied as a lyophilized powder. The preparation is endotoxin-low and biologically active for in vitro antiviral and immunomodulatory assays, intended for research and cell culture applications.

  • Species: human.
  • Expression system: E. coli.
  • Tag: C-terminal 6xHis.
  • Formulation: lyophilized from PBS (pH 8.0) with trehalose.
  • Purity: >95% by reducing SDS-PAGE.
  • Predicted molecular mass: 20.6 kDa (≈20 kDa by SDS-PAGE).
  • Endotoxin: <0.1 EU per μg.
  • Biological activity: ED50 <20 ng/mL in protection assay.
  • Recommended use: for research and cell culture only.
  • Storage: store at -20°C; reconstituted material stable at 4°C for 1 week.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.