Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Interferon lambda 2 (IL-28A), recombinant mouse, His-tagged | >95.0% | 2UG
SDP

Catalog No. 5000177782
Encompass_Preferred

Recombinant mouse interferon-lambda 2 (IL-28A) supplied as a C-terminal His-tagged protein produced in E. coli and provided lyophilized from PBS with trehalose. The product is intended for cell-culture research, has >95% purity by reducing SDS-PAGE, and an apparent molecular weight of approximately 17-25 kDa. Store frozen and aliquot after reconstitution for extended stability.

  • Produced in E. coli and C-terminal His-tagged.
  • Lyophilized from 0.22 μm filtered PBS (pH 7.4) with trehalose.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Apparent molecular weight ~17-25 kDa.
  • For cell culture use only.
  • Store at -20°C; after reconstitution stable 4°C for one week or frozen for longer.

Catalog No. 50-001-77782 Supplier Medchemexpress LLC Supplier No. HYP700203AF2UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse interferon-lambda 2 (IL-28A) supplied as a C-terminal His-tagged protein produced in E. coli and provided lyophilized from PBS with trehalose. The product is intended for cell-culture research, has >95% purity by reducing SDS-PAGE, and an apparent molecular weight of approximately 17-25 kDa. Store frozen and aliquot after reconstitution for extended stability.

  • Produced in E. coli and C-terminal His-tagged.
  • Lyophilized from 0.22 μm filtered PBS (pH 7.4) with trehalose.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Apparent molecular weight ~17-25 kDa.
  • For cell culture use only.
  • Store at -20°C; after reconstitution stable 4°C for one week or frozen for longer.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.