Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IFN-lambda 3 (IL-28B), recombinant human, C-His tag | 95.0% | 50UG
SDP

Catalog No. 5000178357
Encompass_Preferred

Recombinant human interferon-lambda 3 (IL-28B) is a type-III interferon with antiviral, antitumor, and immunomodulatory activity. This animal-free, recombinant protein is produced in E. coli and supplied with a C-terminal His tag to facilitate detection and purification. It is intended for research applications investigating epithelial antiviral signaling and innate immune responses.

  • Recombinant human IFN-lambda 3 (IL-28B), animal-free production.
  • Expressed in E. coli with a C-terminal His tag for purification.
  • Mature protein length 175 amino acids.
  • Purity 95.0%.
  • Supplied as a 50 μg quantity for research use.

Catalog No. 50-001-78357 Supplier Medchemexpress LLC Supplier No. HYP700120AF50UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human interferon-lambda 3 (IL-28B) is a type-III interferon with antiviral, antitumor, and immunomodulatory activity. This animal-free, recombinant protein is produced in E. coli and supplied with a C-terminal His tag to facilitate detection and purification. It is intended for research applications investigating epithelial antiviral signaling and innate immune responses.

  • Recombinant human IFN-lambda 3 (IL-28B), animal-free production.
  • Expressed in E. coli with a C-terminal His tag for purification.
  • Mature protein length 175 amino acids.
  • Purity 95.0%.
  • Supplied as a 50 μg quantity for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.