Learn More
Medchemexpress LLC Animal-free IL-1 beta protein, mouse (His) | 10UG
SDP
Catalog No. 5000177815
Encompass_Preferred
Animal-Free IL-1 beta Protein, Mouse (His) is a recombinant mouse interleukin-1 beta (IL-1β) provided in an animal-free production format with a C-terminal His tag. It is supplied in small vial sizes suitable for in vitro research and biochemical characterization; molecular weight is approximately 17-19 kDa by SDS-PAGE.
  • Recombinant mouse IL-1 beta with C-terminal His tag.
  • Animal-free production to reduce animal-derived contaminants.
  • Available in 10 μg and 50 μg sizes for small-scale experiments.
  • Approximate molecular weight 17-19 kDa (reducing conditions).
  • Datasheet and safety data sheet available for handling and storage information.
Catalog No. 50-001-77815 Supplier Medchemexpress LLC Supplier No. HYP700187AF10UG
Animal-Free IL-1 beta Protein, Mouse (His) is a recombinant mouse interleukin-1 beta (IL-1β) provided in an animal-free production format with a C-terminal His tag. It is supplied in small vial sizes suitable for in vitro research and biochemical characterization; molecular weight is approximately 17-19 kDa by SDS-PAGE.
  • Recombinant mouse IL-1 beta with C-terminal His tag.
  • Animal-free production to reduce animal-derived contaminants.
  • Available in 10 μg and 50 μg sizes for small-scale experiments.
  • Approximate molecular weight 17-19 kDa (reducing conditions).
  • Datasheet and safety data sheet available for handling and storage information.