Learn More
Medchemexpress LLC Animal-free IL-13 protein, mouse (His) | 10UG
SDP
Supplier:  Medchemexpress LLC HYP700192AF10UG
Encompass_Preferred
Recombinant mouse interleukin-13 (IL-13) produced under animal-free conditions and supplied with a polyhistidine tag for purification and detection. The protein is intended for research use in in vitro assays, including cell signaling and cytokine stimulation studies. SDS/PAGE data indicate an approximate molecular weight of 11 kDa. Pack sizes are suitable for assay-scale experiments.
  • Animal-free preparation reduces risk of animal-derived contaminants.
  • His tag enables efficient purification and detection.
  • Suitable for in vitro cell culture and cytokine stimulation assays.
  • Approximate molecular weight 11.0 kDa (SDS-PAGE) aids experimental planning.
  • Pack sizes appropriate for assay-scale experiments.
Catalog No. 50-001-78023