Learn More
Medchemexpress LLC Animal-free IL-13 protein, mouse (His) | 2UG
SDP
Catalog No. 5000177547
Encompass_Preferred
Recombinant mouse interleukin-13 (IL-13) is expressed in E. coli and supplied as a His-tagged, animal-free lyophilized powder for research use. It is intended for cell-based assays and bioactivity testing, with documented activity in TF-1 cell proliferation assays.
  • His-tagged protein for detection and purification.
  • Animal-free production suitable for cell culture applications.
  • Lyophilized powder; recommended reconstitution to 100-200 μg/mL in ddH2O.
  • Approximate molecular weight ~11 kDa as determined by SDS-PAGE.
  • High specific activity (>2.5x10^5 IU/mg); ED50 <10 ng/mL in TF-1 assay.
  • Store at -20°C for long-term storage; add carrier protein for stability after reconstitution.
Catalog No. 50-001-77547 Supplier Medchemexpress LLC Supplier No. HYP700192AF2UG
Recombinant mouse interleukin-13 (IL-13) is expressed in E. coli and supplied as a His-tagged, animal-free lyophilized powder for research use. It is intended for cell-based assays and bioactivity testing, with documented activity in TF-1 cell proliferation assays.
  • His-tagged protein for detection and purification.
  • Animal-free production suitable for cell culture applications.
  • Lyophilized powder; recommended reconstitution to 100-200 μg/mL in ddH2O.
  • Approximate molecular weight ~11 kDa as determined by SDS-PAGE.
  • High specific activity (>2.5x10^5 IU/mg); ED50 <10 ng/mL in TF-1 assay.
  • Store at -20°C for long-term storage; add carrier protein for stability after reconstitution.