Learn More
Medchemexpress LLC Interleukin-13 (mouse) | 50UG
SDP
Catalog No. 5000178026
Encompass_Preferred
Recombinant animal-free mouse interleukin-13 with a C-terminal His tag, expressed in E. coli and supplied lyophilized. The preparation is endotoxin-controlled and biologically active, suitable for cell-based assays (for example, TF-1 proliferation) and biochemical studies that require low-endotoxin cytokine preparations.
  • Animal-free recombinant protein suitable for in vitro assays.
  • His-tag for easy purification and detection.
  • Low endotoxin (<0.1 EU/μg) for sensitive applications.
  • Biologically active: ED50 <10 ng/mL in TF-1 proliferation assay.
  • High specific activity (>2.5x10^5 IU/mg).
  • Lyophilized from PBS for stability and handling convenience.
  • Approximate molecular weight ~11 kDa.
  • Recommended reconstitution at 100-200 μg/mL; store at -20°C.
Catalog No. 50-001-78026 Supplier Medchemexpress LLC Supplier No. HYP700192AF50UG
Recombinant animal-free mouse interleukin-13 with a C-terminal His tag, expressed in E. coli and supplied lyophilized. The preparation is endotoxin-controlled and biologically active, suitable for cell-based assays (for example, TF-1 proliferation) and biochemical studies that require low-endotoxin cytokine preparations.
  • Animal-free recombinant protein suitable for in vitro assays.
  • His-tag for easy purification and detection.
  • Low endotoxin (<0.1 EU/μg) for sensitive applications.
  • Biologically active: ED50 <10 ng/mL in TF-1 proliferation assay.
  • High specific activity (>2.5x10^5 IU/mg).
  • Lyophilized from PBS for stability and handling convenience.
  • Approximate molecular weight ~11 kDa.
  • Recommended reconstitution at 100-200 μg/mL; store at -20°C.