Learn More
Medchemexpress LLC Animal-Free IL-15 H 50ug
SDP
Catalog No. 5000179760
Encompass_Preferred
IL-15 protein regulates immune cells stimulates the proliferation of NK cells T cells and B cells and promotes cytokine secretion Animal-Free IL-15 Protein Human (His) is the recombinant human-derived animal-FreeIL-15 protein expressed by E coli with N-His N-His labeled tag This product is for cell culture use only
Catalog No. 50-001-79760 Supplier Medchemexpress LLC Supplier No. HYP7371AF50UG
IL-15 protein regulates immune cells stimulates the proliferation of NK cells T cells and B cells and promotes cytokine secretion Animal-Free IL-15 Protein Human (His) is the recombinant human-derived animal-FreeIL-15 protein expressed by E coli with N-His N-His labeled tag This product is for cell culture use only