Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Interleukin-15, recombinant porcine (His tag) | 10UG
SDP

Catalog No. 5000178162
Encompass_Preferred

Recombinant porcine interleukin-15 (IL-15), His-tagged and produced in E. coli, supplied as an animal-free protein for research use. IL-15 is a cytokine involved in the activation and proliferation of natural killer cells and T lymphocytes, and is suitable for immunology and cell-based assays.

  • Recombinant porcine IL-15 with His tag.
  • Expressed in E. coli for consistent production.
  • Low endotoxin level (<0.01 EU/μg) for sensitive assays.
  • Formulated in 1X PBS, pH 7.4 for buffer compatibility.
  • Approximate molecular weight 11-17 kDa by SDS-PAGE.
  • Recommended storage: aliquot and freeze at -20°C or -80°C for extended storage.

Catalog No. 50-001-78162 Supplier Medchemexpress LLC Supplier No. HYP700245AF10UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant porcine interleukin-15 (IL-15), His-tagged and produced in E. coli, supplied as an animal-free protein for research use. IL-15 is a cytokine involved in the activation and proliferation of natural killer cells and T lymphocytes, and is suitable for immunology and cell-based assays.

  • Recombinant porcine IL-15 with His tag.
  • Expressed in E. coli for consistent production.
  • Low endotoxin level (<0.01 EU/μg) for sensitive assays.
  • Formulated in 1X PBS, pH 7.4 for buffer compatibility.
  • Approximate molecular weight 11-17 kDa by SDS-PAGE.
  • Recommended storage: aliquot and freeze at -20°C or -80°C for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.