Learn More
Medchemexpress LLC Interleukin-15 (porcine), recombinant, His-tag | ≥95.0% | 50UG

Supplier: Medchemexpress LLC HYP700245AF50UG
Recombinant porcine interleukin-15 (N-terminal His-tag), expressed in E. coli and supplied as a lyophilized powder for research use. The product corresponds to accession Q95253 (T49-S162) and is characterized for biological activity and low endotoxin to support cell-based assays.
- Recombinant porcine interleukin-15 with N-terminal His-tag.
- Expressed in E. coli and lyophilized for stability.
- Low endotoxin level (<0.01 EU/μg) suitable for cell assays.
- Biologically active; ED50 = 3.87 ng/mL in TF-1 proliferation assay.
- Approximate molecular weight 11-17 kDa (SDS-PAGE, reducing).
- Store at -20°C; reconstitute to 100-200 μg/mL and aliquot for long-term storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.