Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Porcine interleukin-2, recombinant (C-terminal 6xHis) | >95.0% | 10UG
SDP

Catalog No. 5000177421
Encompass_Preferred

Recombinant porcine interleukin-2 (IL-2) is supplied as an animal-free, C-terminal His-tagged protein expressed in Escherichia coli and provided lyophilized for research use in cell culture and cancer immunotherapy studies.

  • Species: pig
  • Expression host: Escherichia coli
  • Tag: C-terminal 6xHis
  • Formulation: lyophilized from PBS (pH 7.4) with trehalose
  • Recommended reconstitution: 100-200 μg/mL in ddH2O
  • Amount: 10 μg
  • Purity: >95% by reducing SDS-PAGE
  • Storage: store at -20°C; after reconstitution stable 4°C for 1 week or -20°C for longer
  • Applications: cell culture and cancer immunotherapy research

Catalog No. 50-001-77421 Supplier Medchemexpress LLC Supplier No. HYP700247AF10UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant porcine interleukin-2 (IL-2) is supplied as an animal-free, C-terminal His-tagged protein expressed in Escherichia coli and provided lyophilized for research use in cell culture and cancer immunotherapy studies.

  • Species: pig
  • Expression host: Escherichia coli
  • Tag: C-terminal 6xHis
  • Formulation: lyophilized from PBS (pH 7.4) with trehalose
  • Recommended reconstitution: 100-200 μg/mL in ddH2O
  • Amount: 10 μg
  • Purity: >95% by reducing SDS-PAGE
  • Storage: store at -20°C; after reconstitution stable 4°C for 1 week or -20°C for longer
  • Applications: cell culture and cancer immunotherapy research

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.