Learn More
Medchemexpress LLC Interleukin-20 (mouse), recombinant, C-terminal His tag | >95.0% | 50UG
SDP
Catalog No. 5000177343
Encompass_Preferred
Animal-Free IL-20 Protein, Mouse (His) is a recombinant, C-terminal His-tagged full-length mouse interleukin-20 produced in E. coli and provided as a purified protein for in vitro research. Typical quality attributes include ≥95% purity (reducing SDS-PAGE), ~18.52 kDa molecular weight, and a length of 152 amino acids. Store frozen at -20 °C.
  • Recombinant full-length mouse interleukin-20 with C-terminal His tag.
  • High purity (≥95%) as determined by reducing SDS-PAGE.
  • Expressed in E. coli for scalable production.
  • Approximate molecular weight ~18.52 kDa and 152 amino acids.
  • Supplied frozen; store at -20 °C for stability.
Catalog No. 50-001-77343 Supplier Medchemexpress LLC Supplier No. HYP700198AF50UG
Animal-Free IL-20 Protein, Mouse (His) is a recombinant, C-terminal His-tagged full-length mouse interleukin-20 produced in E. coli and provided as a purified protein for in vitro research. Typical quality attributes include ≥95% purity (reducing SDS-PAGE), ~18.52 kDa molecular weight, and a length of 152 amino acids. Store frozen at -20 °C.
  • Recombinant full-length mouse interleukin-20 with C-terminal His tag.
  • High purity (≥95%) as determined by reducing SDS-PAGE.
  • Expressed in E. coli for scalable production.
  • Approximate molecular weight ~18.52 kDa and 152 amino acids.
  • Supplied frozen; store at -20 °C for stability.