Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Recombinant human interleukin-21 (GMP grade) | >95.0% | 50UG
SDP

Catalog No. 5000178325
Encompass_Preferred

Recombinant human interleukin-21 (IL-21) is supplied as a C-terminal His-tagged recombinant protein expressed in E. coli and provided as a lyophilized powder for research use in cell culture and related assays. The product shows high purity by reducing SDS-PAGE and is stored at -20 °C for stability.

  • Recombinant human IL-21 with C-terminal His tag.
  • Expressed in Escherichia coli for scalable production.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Predicted molecular weight 16.2 kDa; observed ~20 kDa by SDS-PAGE.
  • Corresponds to human IL-21 residues 32-162.
  • Supplied lyophilized and suitable for cell culture use only.
  • Store at -20 °C to maintain stability.
  • Available in small research sizes such as 50 μg.

Catalog No. 50-001-78325 Supplier Medchemexpress LLC Supplier No. HYP700112AF50UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human interleukin-21 (IL-21) is supplied as a C-terminal His-tagged recombinant protein expressed in E. coli and provided as a lyophilized powder for research use in cell culture and related assays. The product shows high purity by reducing SDS-PAGE and is stored at -20 °C for stability.

  • Recombinant human IL-21 with C-terminal His tag.
  • Expressed in Escherichia coli for scalable production.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Predicted molecular weight 16.2 kDa; observed ~20 kDa by SDS-PAGE.
  • Corresponds to human IL-21 residues 32-162.
  • Supplied lyophilized and suitable for cell culture use only.
  • Store at -20 °C to maintain stability.
  • Available in small research sizes such as 50 μg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.