Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Interleukin-21 (mouse), recombinant, His-tagged | >95.0% | 10UG
SDP

Catalog No. 5000177486
Encompass_Preferred

Animal-Free IL-21 Protein, Mouse (His) is a recombinant murine interleukin-21 produced in E. coli and supplied as a His-tagged, lyophilized protein for research applications in immunology and cytokine signaling. It offers low endotoxin and high specific activity for reliable bioassays.

  • Recombinant mouse IL-21 covering residues H18-S146.
  • His-tagged for easy detection and purification.
  • Expressed in E. coli; non-glycosylated form.
  • Purity >95% by reducing SDS-PAGE.
  • Endotoxin <0.1 EU per μg (LAL).
  • High biological activity: specific activity >1.6x10^5 IU/mg; ED50 <6 ng/mL.
  • Lyophilized from PBS with trehalose for stability.
  • Store at -20°C; reconstitute and aliquot to avoid freeze-thaw.

Catalog No. 50-001-75923 Supplier Medchemexpress LLC Supplier No. HYP700199AF10UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Animal-Free IL-21 Protein, Mouse (His) is a recombinant murine interleukin-21 produced in E. coli and supplied as a His-tagged, lyophilized protein for research applications in immunology and cytokine signaling. It offers low endotoxin and high specific activity for reliable bioassays.

  • Recombinant mouse IL-21 covering residues H18-S146.
  • His-tagged for easy detection and purification.
  • Expressed in E. coli; non-glycosylated form.
  • Purity >95% by reducing SDS-PAGE.
  • Endotoxin <0.1 EU per μg (LAL).
  • High biological activity: specific activity >1.6x10^5 IU/mg; ED50 <6 ng/mL.
  • Lyophilized from PBS with trehalose for stability.
  • Store at -20°C; reconstitute and aliquot to avoid freeze-thaw.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.