Learn More
Medchemexpress LLC Recombinant mouse interleukin-21 (His-tag) | 95.0% | 2UG
SDP
Catalog No. 5000178312
Encompass_Preferred
Recombinant mouse interleukin-21 (His-tag) is a lyophilized cytokine produced in E. coli for research applications. The product contains the mature IL-21 sequence, shows activity in immune cell assays, and is supplied with low endotoxin to support sensitive biological studies. It is formulated for stability and recommended to be aliquoted and frozen for long-term storage.
  • Recombinant, His-tagged mature mouse IL-21.
  • Expressed in E. coli and supplied as lyophilized powder.
  • Biological activity: ED50 <6 ng/mL in IFN-γ secretion assay.
  • Low endotoxin: <0.1 EU per μg (LAL method).
  • Approximate molecular weight ~16.8 kDa by sequence.
  • Formulated in PBS with trehalose; freeze aliquots for storage.
Catalog No. 50-001-78312 Supplier Medchemexpress LLC Supplier No. HYP700199AF2UG
Recombinant mouse interleukin-21 (His-tag) is a lyophilized cytokine produced in E. coli for research applications. The product contains the mature IL-21 sequence, shows activity in immune cell assays, and is supplied with low endotoxin to support sensitive biological studies. It is formulated for stability and recommended to be aliquoted and frozen for long-term storage.
  • Recombinant, His-tagged mature mouse IL-21.
  • Expressed in E. coli and supplied as lyophilized powder.
  • Biological activity: ED50 <6 ng/mL in IFN-γ secretion assay.
  • Low endotoxin: <0.1 EU per μg (LAL method).
  • Approximate molecular weight ~16.8 kDa by sequence.
  • Formulated in PBS with trehalose; freeze aliquots for storage.