Learn More
Medchemexpress LLC Animal-Free IL-22 M 10ug
SDP
Catalog No. 5000197015
Encompass_Preferred
Recombinant animal-free mouse interleukin-22 (IL-22) with a C-terminal His tag, expressed in E. coli and supplied for cell culture applications. The product corresponds to mouse IL-22 (sequence L34-V179, accession Q9JJY9), has an approximate molecular weight of 17 kDa by SDS-PAGE, and is provided at ~95% purity. Available in small research sizes (2 μg, 10 μg, 50 μg).
  • Animal-free recombinant formulation suitable for cell culture use.
  • C-terminal His tag enables detection and affinity purification.
  • Expressed in E. coli for consistent, scalable production.
  • Approximately 17 kDa molecular weight by SDS-PAGE.
  • High purity (~95%) for reproducible experimental results.
  • Available in small, convenient microgram pack sizes for screening.
Catalog No. 50-001-97015 Supplier Medchemexpress LLC Supplier No. HYP7079AF10UG
Recombinant animal-free mouse interleukin-22 (IL-22) with a C-terminal His tag, expressed in E. coli and supplied for cell culture applications. The product corresponds to mouse IL-22 (sequence L34-V179, accession Q9JJY9), has an approximate molecular weight of 17 kDa by SDS-PAGE, and is provided at ~95% purity. Available in small research sizes (2 μg, 10 μg, 50 μg).
  • Animal-free recombinant formulation suitable for cell culture use.
  • C-terminal His tag enables detection and affinity purification.
  • Expressed in E. coli for consistent, scalable production.
  • Approximately 17 kDa molecular weight by SDS-PAGE.
  • High purity (~95%) for reproducible experimental results.
  • Available in small, convenient microgram pack sizes for screening.