Learn More
Medchemexpress LLC Interleukin-24 (IL-24) | >95.0% | 10UG
SDP
Catalog No. 5000178134
Encompass_Preferred
Animal-Free IL-24 Protein, Mouse (His) is a recombinant mouse interleukin-24 expressed in Escherichia coli and supplied lyophilized with a C-terminal 6xHis tag for research use in cell culture and in vitro assays. The product is highly pure and biologically active, with characterization by SDS-PAGE and functional ED50 data.
Catalog No. 50-001-78134 Supplier Medchemexpress LLC Supplier No. HYP700200AF10UG
Animal-Free IL-24 Protein, Mouse (His) is a recombinant mouse interleukin-24 expressed in Escherichia coli and supplied lyophilized with a C-terminal 6xHis tag for research use in cell culture and in vitro assays. The product is highly pure and biologically active, with characterization by SDS-PAGE and functional ED50 data.