Learn More
Medchemexpress LLC Animal-free IL-24 protein, mouse (His) | >95.0% | 50UG
SDP
Catalog No. 5000178059
Encompass_Preferred
Recombinant mouse interleukin-24 (IL-24), C-terminal His-tagged and expressed in E. coli. Supplied lyophilized from PBS for in vitro cell culture and receptor-mediated bioactivity assays, with high purity and documented potency.
Catalog No. 50-001-78059 Supplier Medchemexpress LLC Supplier No. HYP700200AF50UG
Recombinant mouse interleukin-24 (IL-24), C-terminal His-tagged and expressed in E. coli. Supplied lyophilized from PBS for in vitro cell culture and receptor-mediated bioactivity assays, with high purity and documented potency.