Learn More
Medchemexpress LLC Recombinant human interleukin-26 (IL-26) | 95.0% | 10UG

Supplier: Medchemexpress LLC HYP700117AF10UG
Recombinant human interleukin-26 (IL-26) is an E. coli-expressed cytokine (residues K22-Q171) supplied with a C-terminal His tag for research use. Provided as a 10 μg quantity with ~95.0% purity, the protein is intended for in vitro applications including signaling studies, cytokine stimulation assays, and biochemical characterization.
- Human sequence (residues K22-Q171) corresponding to accession Q9NPH9.
- Expressed in E. coli with a C-terminal His tag for purification.
- Approximately 95.0% purity suitable for biochemical assays.
- Supplied as a 10 μg research-use quantity.
- Molecular weight approximately 18.0 kDa.
- Suitable for in vitro signaling and immune-response studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.