Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Interleukin-31 (mouse) | >95.0% | 10UG
SDP

Catalog No. 5000177783
Encompass_Preferred

Animal-free IL-31 protein is a recombinant, His-tagged mouse interleukin-31 produced in Escherichia coli and supplied lyophilized. It corresponds to residues T24-C163 (accession Q6EAL8), has a predicted molecular weight of 16.6 kDa (observed ≈17.0 kDa), and is >95% pure by reducing SDS-PAGE. Formulated in PBS (pH 7.4) with trehalose, it is intended for research use and stored at -20°C.

  • Recombinant His-tagged mouse interleukin-31
  • Expressed in Escherichia coli
  • Sequence range T24-C163 (accession Q6EAL8)
  • Predicted 16.6 kDa; observed ≈17.0 kDa
  • Purity greater than 95% by reducing SDS-PAGE
  • Lyophilized formulation in PBS pH 7.4 with trehalose
  • Store at -20°C; freeze aliquots for long-term storage

Catalog No. 50-001-77783 Supplier Medchemexpress LLC Supplier No. HYP700207AF10UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Animal-free IL-31 protein is a recombinant, His-tagged mouse interleukin-31 produced in Escherichia coli and supplied lyophilized. It corresponds to residues T24-C163 (accession Q6EAL8), has a predicted molecular weight of 16.6 kDa (observed ≈17.0 kDa), and is >95% pure by reducing SDS-PAGE. Formulated in PBS (pH 7.4) with trehalose, it is intended for research use and stored at -20°C.

  • Recombinant His-tagged mouse interleukin-31
  • Expressed in Escherichia coli
  • Sequence range T24-C163 (accession Q6EAL8)
  • Predicted 16.6 kDa; observed ≈17.0 kDa
  • Purity greater than 95% by reducing SDS-PAGE
  • Lyophilized formulation in PBS pH 7.4 with trehalose
  • Store at -20°C; freeze aliquots for long-term storage

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.