Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Animal-free interleukin-31 protein (mouse, His-tag) | >95.0% | 50UG
SDP

Catalog No. 5000178274
Encompass_Preferred

Recombinant mouse interleukin-31 (IL-31), produced in an animal-free Escherichia coli expression system and C-terminally His-tagged, supplied for cell culture and research applications investigating IL-31 signaling pathways.

  • Species: mouse.
  • Expression system: Escherichia coli.
  • Tag: C-His (C-terminal).
  • Purity: >95.0% as determined by reducing SDS-PAGE.
  • Predicted molecular weight: 16.6 kDa.
  • Sequence/residue range: T24-C163.
  • Biological activity: activates STAT3; may activate STAT1 and STAT5.
  • Intended use: for cell culture use only.

Catalog No. 50-001-78274 Supplier Medchemexpress LLC Supplier No. HYP700207AF50UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse interleukin-31 (IL-31), produced in an animal-free Escherichia coli expression system and C-terminally His-tagged, supplied for cell culture and research applications investigating IL-31 signaling pathways.

  • Species: mouse.
  • Expression system: Escherichia coli.
  • Tag: C-His (C-terminal).
  • Purity: >95.0% as determined by reducing SDS-PAGE.
  • Predicted molecular weight: 16.6 kDa.
  • Sequence/residue range: T24-C163.
  • Biological activity: activates STAT3; may activate STAT1 and STAT5.
  • Intended use: for cell culture use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.