Learn More
Medchemexpress LLC Animal-Free IL-33 H 10ug
SDP
Catalog No. 5000185498
Encompass_Preferred
Animal-Free IL-33 Protein, Human (His) is a recombinant His-tagged fragment of human interleukin-33 (residues S112-T270) produced using an animal-free manufacturing process for in vitro research. It is supplied in research-scale sizes suitable for signaling and receptor interaction studies.
  • Recombinant human IL-33 fragment (S112-T270) suitable for in vitro assays.
  • C-terminal 6xHis tag for detection and purification.
  • Produced under animal-free manufacturing conditions for high consistency.
  • Available in small research sizes, including 10 μg.
  • Intended for cell signaling, receptor binding, and functional cytokine assays.
Catalog No. 50-001-85498 Supplier Medchemexpress LLC Supplier No. HYP70475AF10UG
Animal-Free IL-33 Protein, Human (His) is a recombinant His-tagged fragment of human interleukin-33 (residues S112-T270) produced using an animal-free manufacturing process for in vitro research. It is supplied in research-scale sizes suitable for signaling and receptor interaction studies.
  • Recombinant human IL-33 fragment (S112-T270) suitable for in vitro assays.
  • C-terminal 6xHis tag for detection and purification.
  • Produced under animal-free manufacturing conditions for high consistency.
  • Available in small research sizes, including 10 μg.
  • Intended for cell signaling, receptor binding, and functional cytokine assays.