Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Interleukin-36 gamma (IL-1F9) | >95.0% | 2UG
SDP

Catalog No. 5000177400
Encompass_Preferred

Recombinant, animal-free mouse interleukin-36 gamma (IL-1F9) expressed in E. coli with a C-terminal 6xHis tag. It activates NF-κB signaling via IL1RL2 and promotes inflammatory responses in epithelial tissues by influencing keratinocytes, dendritic cells, and T cells. Supplied lyophilized for research use in cell culture.

  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Predicted molecular mass 18.3 kDa; ~17 kDa on SDS-PAGE.
  • Contains sequence corresponding to G13-S164 of mouse IL-36γ.
  • Lyophilized formulation in PBS (pH 7.4) with trehalose for stability.
  • Reconstitute to 100-200 μg/mL in ddH2O for typical use.
  • Stable frozen at -20 °C for up to 2 years; aliquot for long-term storage.
  • Intended for cell culture research use only.

Catalog No. 50-001-77400 Supplier Medchemexpress LLC Supplier No. HYP700211AF2UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant, animal-free mouse interleukin-36 gamma (IL-1F9) expressed in E. coli with a C-terminal 6xHis tag. It activates NF-κB signaling via IL1RL2 and promotes inflammatory responses in epithelial tissues by influencing keratinocytes, dendritic cells, and T cells. Supplied lyophilized for research use in cell culture.

  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Predicted molecular mass 18.3 kDa; ~17 kDa on SDS-PAGE.
  • Contains sequence corresponding to G13-S164 of mouse IL-36γ.
  • Lyophilized formulation in PBS (pH 7.4) with trehalose for stability.
  • Reconstitute to 100-200 μg/mL in ddH2O for typical use.
  • Stable frozen at -20 °C for up to 2 years; aliquot for long-term storage.
  • Intended for cell culture research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.