Learn More
Medchemexpress LLC Animal-free IL-36 gamma/IL-1F9 protein, mouse (His) | >95.0% | 50UG
SDP
Catalog No. 5000178184
Encompass_Preferred
Recombinant animal-free mouse interleukin-36 gamma (IL-36γ/IL-1F9) supplied with a His tag. The protein corresponds to residues G13-S164 and is expressed in E. coli. It is purified for use in research on IL-36 signaling, inflammation, and related cellular assays. The product is provided in multiple sizes, stored frozen, and includes manufacturer handling and storage recommendations.
Catalog No. 50-001-78184 Supplier Medchemexpress LLC Supplier No. HYP700211AF50UG
Recombinant animal-free mouse interleukin-36 gamma (IL-36γ/IL-1F9) supplied with a His tag. The protein corresponds to residues G13-S164 and is expressed in E. coli. It is purified for use in research on IL-36 signaling, inflammation, and related cellular assays. The product is provided in multiple sizes, stored frozen, and includes manufacturer handling and storage recommendations.