Learn More
Medchemexpress LLC Animal-free IL-37 protein, human (His) | >95.0% | 50UG
SDP
Catalog No. 5000178186
Encompass_Preferred
Recombinant human interleukin-37 (IL-37) with a C-terminal 6xHis tag, expressed in Escherichia coli and supplied as a lyophilized powder. The protein is high purity and biologically active for in vitro immunology and signaling studies, formulated in PBS (pH 8.0) with trehalose, with low endotoxin and recommended storage at -20°C. Reconstitute to 100-200 μg/mL in ddH2O and aliquot for long-term storage.
Catalog No. 50-001-78186 Supplier Medchemexpress LLC Supplier No. HYP700128AF50UG
Recombinant human interleukin-37 (IL-37) with a C-terminal 6xHis tag, expressed in Escherichia coli and supplied as a lyophilized powder. The protein is high purity and biologically active for in vitro immunology and signaling studies, formulated in PBS (pH 8.0) with trehalose, with low endotoxin and recommended storage at -20°C. Reconstitute to 100-200 μg/mL in ddH2O and aliquot for long-term storage.