Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Animal-free IL-4 protein, pig (His) | 98.0% | 50UG
SDP

Catalog No. 5000177790
Encompass_Preferred

Recombinant porcine interleukin-4 (IL-4) supplied lyophilized and produced in E. coli. The protein contains a C-terminal 6xHis tag, has an approximate molecular weight of 15 kDa, and is ≥98.0% pure by reducing SDS-PAGE. Biological activity is validated (ED50 <0.6 ng/mL); reconstitute to 100-200 μg/mL and store lyophilized at -20 °C or aliquot and freeze at -20 °C/-80 °C for long-term storage.

  • Recombinant porcine interleukin-4 in lyophilized form.
  • C-terminal 6xHis tag for detection or purification.
  • Produced in E. coli expression system.
  • Approximate molecular weight of 15 kDa.
  • Purity of 98.0% as determined by SDS-PAGE.
  • Biologically active with ED50 <0.6 ng/mL in TF-1 assay.
  • Formulated from 0.22 μm filtered PBS, pH 7.4.
  • Stable lyophilized at -20 °C; aliquot and store at -20 °C/-80 °C after reconstitution.

Catalog No. 50-001-77790 Supplier Medchemexpress LLC Supplier No. HYP700248AF50UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant porcine interleukin-4 (IL-4) supplied lyophilized and produced in E. coli. The protein contains a C-terminal 6xHis tag, has an approximate molecular weight of 15 kDa, and is ≥98.0% pure by reducing SDS-PAGE. Biological activity is validated (ED50 <0.6 ng/mL); reconstitute to 100-200 μg/mL and store lyophilized at -20 °C or aliquot and freeze at -20 °C/-80 °C for long-term storage.

  • Recombinant porcine interleukin-4 in lyophilized form.
  • C-terminal 6xHis tag for detection or purification.
  • Produced in E. coli expression system.
  • Approximate molecular weight of 15 kDa.
  • Purity of 98.0% as determined by SDS-PAGE.
  • Biologically active with ED50 <0.6 ng/mL in TF-1 assay.
  • Formulated from 0.22 μm filtered PBS, pH 7.4.
  • Stable lyophilized at -20 °C; aliquot and store at -20 °C/-80 °C after reconstitution.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.