Learn More
Medchemexpress LLC Recombinant mouse interleukin-7/IL-7 protein (His tag) | 95.0% | 10UG
SDP
Catalog No. 5000177785
Encompass_Preferred
Recombinant mouse interleukin-7 (IL-7) produced in Escherichia coli with a C-terminal His tag. This animal-free recombinant cytokine promotes lymphocyte development and survival by engaging the IL7RA/CSF2RG receptor complex and activating downstream signaling pathways (JAK/STAT, PI3K/Akt). Intended for research cell culture applications, it is supplied as a small vial and should be stored at -20°C.
Catalog No. 50-001-77785 Supplier Medchemexpress LLC Supplier No. HYP700217AF10UG
Recombinant mouse interleukin-7 (IL-7) produced in Escherichia coli with a C-terminal His tag. This animal-free recombinant cytokine promotes lymphocyte development and survival by engaging the IL7RA/CSF2RG receptor complex and activating downstream signaling pathways (JAK/STAT, PI3K/Akt). Intended for research cell culture applications, it is supplied as a small vial and should be stored at -20°C.