Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mouse interleukin-7, recombinant, His-tagged | 95.0% | 50UG
SDP

Catalog No. 5000177786
Encompass_Preferred

Animal-Free IL-7 Protein, Mouse (His) is a recombinant, His-tagged mouse interleukin-7 produced using an animal-free workflow and supplied as a research reagent for immunology and cell-culture applications. It is intended for in vitro studies of lymphocyte development, survival, and signaling.

  • Recombinant mouse interleukin-7 produced in E. coli.
  • His-tagged for detection and purification.
  • Animal-free production suitable for cell-culture applications.
  • Approximately 16.0 kDa molecular weight.
  • Purity ≥95.0% by manufacturer specification.
  • Provided as a 50 μg reagent for in vitro assays.

Catalog No. 50-001-77786 Supplier Medchemexpress LLC Supplier No. HYP700217AF50UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Animal-Free IL-7 Protein, Mouse (His) is a recombinant, His-tagged mouse interleukin-7 produced using an animal-free workflow and supplied as a research reagent for immunology and cell-culture applications. It is intended for in vitro studies of lymphocyte development, survival, and signaling.

  • Recombinant mouse interleukin-7 produced in E. coli.
  • His-tagged for detection and purification.
  • Animal-free production suitable for cell-culture applications.
  • Approximately 16.0 kDa molecular weight.
  • Purity ≥95.0% by manufacturer specification.
  • Provided as a 50 μg reagent for in vitro assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.