Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Polypropylene, Straight, Male Luer to Hose Barb Adapter, 3/16" ID; 25/PK
Catalog No. NC1338732
Encompass
Manufactured of animal-free natural polypropylene, these luers meet Class VI specifications. Luers offer superior chemical resistance and are suited for ETO and gamma sterilization.
Catalog No. NC1338732 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3080020
Manufactured of animal-free natural polypropylene, these luers meet Class VI specifications. Luers offer superior chemical resistance and are suited for ETO and gamma sterilization.