Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Animal-free NAP-2/CXCL7 protein, human (His) | >95.0% | 2UG
SDP

Catalog No. 5000177499
Encompass_Preferred

Recombinant human NAP-2/CXCL7 protein fragment with an N-terminal 6xHis tag, produced in E. coli and supplied with a certificate of analysis. The preparation corresponds to residues A59-D128 (70 amino acids), has an approximate molecular weight of ~11 kDa, and is ≥95.0% pure by reducing SDS-PAGE. For research use only.

  • Recombinant fragment representing residues A59-D128.
  • N-terminal 6xHis tag for purification and detection.
  • Produced in E. coli for efficient expression.
  • Purity ≥95.0% as determined by reducing SDS-PAGE.
  • Provided with a certificate of analysis for batch-specific data.
  • Suitable for biochemical assays and antibody development.

Catalog No. 50-001-77499 Supplier Medchemexpress LLC Supplier No. HYP700049AF2UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human NAP-2/CXCL7 protein fragment with an N-terminal 6xHis tag, produced in E. coli and supplied with a certificate of analysis. The preparation corresponds to residues A59-D128 (70 amino acids), has an approximate molecular weight of ~11 kDa, and is ≥95.0% pure by reducing SDS-PAGE. For research use only.

  • Recombinant fragment representing residues A59-D128.
  • N-terminal 6xHis tag for purification and detection.
  • Produced in E. coli for efficient expression.
  • Purity ≥95.0% as determined by reducing SDS-PAGE.
  • Provided with a certificate of analysis for batch-specific data.
  • Suitable for biochemical assays and antibody development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.