Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Animal-free SUMO protease protein, Saccharomyces cerevisiae (His) | >95.0% | 1000U
SDP

Catalog No. 5000175392
Encompass_Preferred

Animal-free SUMO protease is a recombinant SUMO-specific protease derived from Saccharomyces cerevisiae and expressed in Escherichia coli. It is supplied as a filtered, glycerol-containing solution with high purity and low endotoxin, intended for efficient cleavage of SUMO fusion tags during protein purification workflows.

  • His-tagged recombinant enzyme for specific SUMO tag cleavage.
  • Biological activity 10 U/μL (one unit cleaves ≥85% of 2 μg substrate in 1 h at 30°C).
  • Formulated in 25 mM Tris-HCl, 0.1% Igepal (NP-40), 250 mM NaCl, 0.5 mM DTT, 50% glycerol, pH 8.0.
  • Filtered 0.22 μm and supplied as a ready-to-use solution.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Endotoxin level <0.2 EU/μg.
  • Store at -20°C; stable 1 year unopened, 3 months after opening if aliquoted.

Catalog No. 50-001-75392 Supplier Medchemexpress LLC Supplier No. HYP701239AF1000U
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Animal-free SUMO protease is a recombinant SUMO-specific protease derived from Saccharomyces cerevisiae and expressed in Escherichia coli. It is supplied as a filtered, glycerol-containing solution with high purity and low endotoxin, intended for efficient cleavage of SUMO fusion tags during protein purification workflows.

  • His-tagged recombinant enzyme for specific SUMO tag cleavage.
  • Biological activity 10 U/μL (one unit cleaves ≥85% of 2 μg substrate in 1 h at 30°C).
  • Formulated in 25 mM Tris-HCl, 0.1% Igepal (NP-40), 250 mM NaCl, 0.5 mM DTT, 50% glycerol, pH 8.0.
  • Filtered 0.22 μm and supplied as a ready-to-use solution.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Endotoxin level <0.2 EU/μg.
  • Store at -20°C; stable 1 year unopened, 3 months after opening if aliquoted.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.