Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Animal-free TGF beta 1 (TGFB1), pig (His tag) | 10UG
SDP

Catalog No. 5000178277
Encompass_Preferred

Recombinant His-tagged porcine transforming growth factor beta 1 (TGF-β1) supplied as an animal-free, lyophilized research reagent. Expressed in E. coli and formulated for stability, it is intended for cell signaling and growth-factor assays.

  • Species: pig.
  • Tag: His tag.
  • Source: expressed in E. coli.
  • Formulation: lyophilized from 20 mM sodium citrate, 0.2 M NaCl, pH 4.5, with trehalose.
  • Endotoxin level: <0.1 EU per 1 μg.
  • Molecular weight: approximately 11-17 kDa by SDS-PAGE.
  • Storage: store at -20°C; after reconstitution stable at 4°C for 1 week or -20°C for longer (aliquot and freeze).
  • Typical packaging: small lyophilized vials (for example, 10 μg).

Catalog No. 50-001-78277 Supplier Medchemexpress LLC Supplier No. HYP700251AF10UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant His-tagged porcine transforming growth factor beta 1 (TGF-β1) supplied as an animal-free, lyophilized research reagent. Expressed in E. coli and formulated for stability, it is intended for cell signaling and growth-factor assays.

  • Species: pig.
  • Tag: His tag.
  • Source: expressed in E. coli.
  • Formulation: lyophilized from 20 mM sodium citrate, 0.2 M NaCl, pH 4.5, with trehalose.
  • Endotoxin level: <0.1 EU per 1 μg.
  • Molecular weight: approximately 11-17 kDa by SDS-PAGE.
  • Storage: store at -20°C; after reconstitution stable at 4°C for 1 week or -20°C for longer (aliquot and freeze).
  • Typical packaging: small lyophilized vials (for example, 10 μg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.