Learn More
Medchemexpress LLC Animal-Free TGF beta 10ug
SDP
Catalog No. 5000177827
Encompass_Preferred
In its proprotein form the TGF beta-1/TGFB1 protein serves as a precursor to latency-associated peptide (LAP) and active transforming growth factor beta-1 (TGF-beta-1) chains Critical to maintaining the latent state of TGF- -1 within the extracellular matrix preprotein interacts with environmental molecules such as LTBP1 LRRC32/GARP and LRRC33/NRROS to regulate TGF- -1 activation Animal-Free TGF beta 1/TGFB1 Protein Mouse/Rat (His) is the recombinant mouse-derived animal-FreeTGF beta 1/TGFB1 protein expressed by E coli with C-His labeled tag This product is for cell culture use only
Catalog No. 50-001-77827 Supplier Medchemexpress LLC Supplier No. HYP700227AF10UG
In its proprotein form the TGF beta-1/TGFB1 protein serves as a precursor to latency-associated peptide (LAP) and active transforming growth factor beta-1 (TGF-beta-1) chains Critical to maintaining the latent state of TGF- -1 within the extracellular matrix preprotein interacts with environmental molecules such as LTBP1 LRRC32/GARP and LRRC33/NRROS to regulate TGF- -1 activation Animal-Free TGF beta 1/TGFB1 Protein Mouse/Rat (His) is the recombinant mouse-derived animal-FreeTGF beta 1/TGFB1 protein expressed by E coli with C-His labeled tag This product is for cell culture use only