Learn More
Medchemexpress LLC Transforming growth factor beta 1 (recombinant pig, His-tagged) | 2UG
SDP
Catalog No. 5000177416
Encompass_Preferred
Recombinant animal-free transforming growth factor beta 1 (TGF-β1) derived from pig, supplied as a His-tagged, lyophilized protein produced in Escherichia coli. The preparation corresponds to the mature chain (A279-S390), demonstrates biological activity (ED50 <0.1 ng/mL in HT-2 assay), and is formulated for stability and long-term storage.
Catalog No. 50-001-77416 Supplier Medchemexpress LLC Supplier No. HYP700251AF2UG
Recombinant animal-free transforming growth factor beta 1 (TGF-β1) derived from pig, supplied as a His-tagged, lyophilized protein produced in Escherichia coli. The preparation corresponds to the mature chain (A279-S390), demonstrates biological activity (ED50 <0.1 ng/mL in HT-2 assay), and is formulated for stability and long-term storage.