Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Human transforming growth factor beta 3 | 67763-96-6 | >95.0% | 50UG
SDP

Catalog No. 5000177590
Encompass_Preferred

Recombinant human transforming growth factor beta-3 (TGF-β3), His-tagged and expressed in E. coli, supplied as a lyophilized powder for in vitro research and bioactivity assays. The protein corresponds to accession P10600-1 (A301-S412) and is formulated for stability and low endotoxin for sensitive cell-based applications.

  • Recombinant human TGF-β3 (mature region A301-S412)
  • His-tagged for detection and purification
  • Expressed in E. coli and supplied lyophilized
  • Purity greater than 95% by reducing SDS-PAGE
  • Endotoxin <0.1 EU per μg as determined by LAL
  • Specific activity >2 x 10^7 IU/mg; ED50 <50 pg/mL in HT-2 assay
  • Recommended reconstitution to 100-200 μg/mL in ddH2O
  • Store lyophilized at -20°C; aliquot and freeze at -80°C for long term

Catalog No. 50-001-77590 Supplier Medchemexpress LLC Supplier No. HYP700152AF50UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant human transforming growth factor beta-3 (TGF-β3), His-tagged and expressed in E. coli, supplied as a lyophilized powder for in vitro research and bioactivity assays. The protein corresponds to accession P10600-1 (A301-S412) and is formulated for stability and low endotoxin for sensitive cell-based applications.

  • Recombinant human TGF-β3 (mature region A301-S412)
  • His-tagged for detection and purification
  • Expressed in E. coli and supplied lyophilized
  • Purity greater than 95% by reducing SDS-PAGE
  • Endotoxin <0.1 EU per μg as determined by LAL
  • Specific activity >2 x 10^7 IU/mg; ED50 <50 pg/mL in HT-2 assay
  • Recommended reconstitution to 100-200 μg/mL in ddH2O
  • Store lyophilized at -20°C; aliquot and freeze at -80°C for long term

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.