Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Animal-free TGF beta 1 (TGFB1) protein, human (His-tag) | >95.0% | 50UG
SDP

Catalog No. 5000177762
Encompass_Preferred

Recombinant human TGF beta 1 (transforming growth factor beta-1) supplied as an animal-free, C-terminal his-tagged, lyophilized protein for research use. Expressed in E. coli and purified to high purity with low endotoxin; recommended reconstitution is 100-200 μg/mL in 10 mM HCl and storage at -20 °C.

  • Animal-free recombinant protein suitable for cell culture and signaling studies.
  • C-terminal his tag enables affinity purification and detection.
  • Expressed in E. coli for consistent lot-to-lot production.
  • Purity greater than 95.0% by reducing SDS-PAGE.
  • Low endotoxin level <0.1 EU per μg for cell-based assays.
  • Lyophilized formulation with recommended reconstitution 100-200 μg/mL in 10 mM HCl.
  • Store at -20 °C; aliquot after reconstitution for long-term stability.

Catalog No. 50-001-77762 Supplier Medchemexpress LLC Supplier No. HYP700150AF50UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human TGF beta 1 (transforming growth factor beta-1) supplied as an animal-free, C-terminal his-tagged, lyophilized protein for research use. Expressed in E. coli and purified to high purity with low endotoxin; recommended reconstitution is 100-200 μg/mL in 10 mM HCl and storage at -20 °C.

  • Animal-free recombinant protein suitable for cell culture and signaling studies.
  • C-terminal his tag enables affinity purification and detection.
  • Expressed in E. coli for consistent lot-to-lot production.
  • Purity greater than 95.0% by reducing SDS-PAGE.
  • Low endotoxin level <0.1 EU per μg for cell-based assays.
  • Lyophilized formulation with recommended reconstitution 100-200 μg/mL in 10 mM HCl.
  • Store at -20 °C; aliquot after reconstitution for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.