Learn More
Medchemexpress LLC Tumor necrosis factor beta, recombinant human (His-tagged) | 95.0% | 50UG
SDP
Catalog No. 5000177754
Encompass_Preferred
Animal-Free TNF-beta (lymphotoxin-alpha) is a recombinant human cytokine produced without animal-derived components. It is expressed in E. coli with an N-terminal His tag, forms biologically active homotrimers, and is intended for in vitro research into TNF signaling and immune responses. Supplied in research quantities (e.g., 50 μg); molecular weight ≈18.65 kDa; purity reported by distributors as ~95%.
  • Recombinant human TNF-beta (lymphotoxin-alpha) for in vitro research.
  • Produced without animal-derived components.
  • Expressed in E. coli and carrying an N-terminal His tag.
  • Forms a biologically relevant homotrimer that binds TNF receptors.
  • Supplied in small research quantities and approximately 18.65 kDa molecular weight.
  • High purity suitable for biochemical and cell-based assays (purity reported as ~95%).
Catalog No. 50-001-77754 Supplier Medchemexpress LLC Supplier No. HYP700154AF50UG
Animal-Free TNF-beta (lymphotoxin-alpha) is a recombinant human cytokine produced without animal-derived components. It is expressed in E. coli with an N-terminal His tag, forms biologically active homotrimers, and is intended for in vitro research into TNF signaling and immune responses. Supplied in research quantities (e.g., 50 μg); molecular weight ≈18.65 kDa; purity reported by distributors as ~95%.
  • Recombinant human TNF-beta (lymphotoxin-alpha) for in vitro research.
  • Produced without animal-derived components.
  • Expressed in E. coli and carrying an N-terminal His tag.
  • Forms a biologically relevant homotrimer that binds TNF receptors.
  • Supplied in small research quantities and approximately 18.65 kDa molecular weight.
  • High purity suitable for biochemical and cell-based assays (purity reported as ~95%).