Learn More
MSC SKILCRAFT Floor Pad, Animal Hair, Polishing, 15 in Diameter, Beige
Catalog No. 502839701
Encompass
Made of curled animal hair, these high-speed pads are designed for use with rotary disk floor maintenance machines. They are 1 inch thick, washable, and can be used on machines operating at up to 2500 rpm. Each box contains 5 pads.
  • Removes scuff marks and light soil
  • Restores high-gloss shine to floors
  • Multi-surface compatibility
  • Washable
Catalog No. 50-283-9701 Supplier MSC Supplier No. 89461057
Made of curled animal hair, these high-speed pads are designed for use with rotary disk floor maintenance machines. They are 1 inch thick, washable, and can be used on machines operating at up to 2500 rpm. Each box contains 5 pads.
  • Removes scuff marks and light soil
  • Restores high-gloss shine to floors
  • Multi-surface compatibility
  • Washable