Learn More
Mercodia Inc Animal Insulin Control, Large Animal, Low Medium High
SDP
Catalog No. NC1720191
Encompass_Preferred
Animal Insulin Control(10-1221-01)Kit format: 3 x 0.5 mLMeasurement range: low, medium, high.Mercodia Animal Insulin Control (Low, Medium, High) is designed to be used as a three-level control for Mercodia Porcine, Bovine, Ovine, Canine and Equine Insulin ELISA kits.
Catalog No. NC1720191 Supplier Mercodia Inc Supplier No. 10122101
Animal Insulin Control(10-1221-01)Kit format: 3 x 0.5 mLMeasurement range: low, medium, high.Mercodia Animal Insulin Control (Low, Medium, High) is designed to be used as a three-level control for Mercodia Porcine, Bovine, Ovine, Canine and Equine Insulin ELISA kits.