Learn More
Supertek™ Animal and Plant Cell Model
GREENER_CHOICESCI_ED
Catalog No. S81672 Shop All Supertek Scientific Products
Change view
Click to view available options
Includes:
Animal cell 3D model and Plant cell magnified model
Catalog No. Includes
S81672 Animal cell 3D model and Plant cell magnified model
1 options
Catalog No. S81672 Supplier Supertek™ Supplier No. B12674ASP
Add to Cart
Add to Cart
Animal Cell and Plant cell models to show internal structures.
  • Animal Cell is a three dimensional model
    • One side dissected to show complete internal structures
    • Mounted on base with key card
  • Plant cell is a highly enlarged model, as seen under electron microscope
    • A portion of the wall is removed to show ectoplast, endoplast, tonoplast, vacuoles, nuclear structure, plastids, mitochondria etc.
    • Mounted on base with key
Specifications
For Use With (Application) Biology and Earth Science
Includes Animal cell 3D model and Plant cell magnified model
Material Vinyl
Product Type Model, Teaching