Learn More
Caron Products And Services Animal Rearing Chamber, 33 cu. ft., LED Lights, Temp, No RH
Catalog No. NC2676815
Encompass_Preferred
Carons 33 cu. ft. (934 liters) Animal Rearing Chamber. Standard features include touchscreen controls, temperature range from 10C to 40C, LED lighting, and light tight door cover. These chambers allow you to simulate specialized environments for your research needs. 115V, 60 Hz.
Catalog No. NC2676815 Supplier Caron Products And Services Supplier No. 7350331
Add to Cart
Add to Cart
Request a Quote
Carons 33 cu. ft. (934 liters) Animal Rearing Chamber. Standard features include touchscreen controls, temperature range from 10C to 40C, LED lighting, and light tight door cover. These chambers allow you to simulate specialized environments for your research needs. 115V, 60 Hz.