Learn More
RWD LIFE SCIENCE INC ANIMAL VENTILATOR
SDP
Catalog No. NC3404314
Encompass_Preferred
R415 is a mechanical ventilator used to assist animal respiration, suitable for mice, rats, and guinea pigs, and other animals weighing 10g-1kg.It is widely used in basic medical research, clinical medicine,animal medicine and other scientific research experiments (such as myocardial ischemia model making, lung function evaluation).
Catalog No. NC3404314 Supplier RWD LIFE SCIENCE INC Supplier No. R415
Add to Cart
Edge
Add to Cart
R415 is a mechanical ventilator used to assist animal respiration, suitable for mice, rats, and guinea pigs, and other animals weighing 10g-1kg.It is widely used in basic medical research, clinical medicine,animal medicine and other scientific research experiments (such as myocardial ischemia model making, lung function evaluation).