Learn More
MSC SKILCRAFT Floor Pad, Animal Hair, Burnishing, Gray, 18 inch Diameter
Catalog No. 502839655
Encompass
This floor pad is made of curled animal hair and designed for high-speed rotary disk floor maintenance machines. It is 1 inch thick, washable, and can be used on machines up to 2500 rpm. Each box contains 5 pads.
  • Made of curled animal hair
  • High-speed pad
  • Used for rotary disk floor maintenance machines
  • 1 inch thick
  • Washable
  • Suitable for machines up to 2500 rpm
  • Gray color
  • 18 inch diameter
Catalog No. 50-283-9655 Supplier MSC Supplier No. 89463137
This floor pad is made of curled animal hair and designed for high-speed rotary disk floor maintenance machines. It is 1 inch thick, washable, and can be used on machines up to 2500 rpm. Each box contains 5 pads.
  • Made of curled animal hair
  • High-speed pad
  • Used for rotary disk floor maintenance machines
  • 1 inch thick
  • Washable
  • Suitable for machines up to 2500 rpm
  • Gray color
  • 18 inch diameter