Learn More
Med Vet International Animalintex Poultice Hoof Shaped Pads. 5 1/2" X 5 1/2" 3/Pack
Catalog No. 502828499
Encompass_Preferred
This hoof poultice is made of 100% cotton wool with a non-woven cover on one side and plastic backing on the other. Contains mild antiseptic (boric acid) plus natural poultice agent (tragacanth). Use as a poultice or wound dressing.- Ideal for cuts, wounds, abscesses, inflammation, skin disorders and hoof conditions- Use as a hot poultice for abscess, boils, infected and dirty wounds, cracked heels, thorns, laminitis, seedy toe, corns- Use as a cold poultice for strains, sprains, sore shins, splints, bruises, capped hocks/elbows- Use as a dry dressing over open wounds or for pressure padding- Pre-cut hoof shape
Catalog No. 50-282-8499 Supplier Med Vet International Supplier No. PHLINTEX3PK
This hoof poultice is made of 100% cotton wool with a non-woven cover on one side and plastic backing on the other. Contains mild antiseptic (boric acid) plus natural poultice agent (tragacanth). Use as a poultice or wound dressing.- Ideal for cuts, wounds, abscesses, inflammation, skin disorders and hoof conditions- Use as a hot poultice for abscess, boils, infected and dirty wounds, cracked heels, thorns, laminitis, seedy toe, corns- Use as a cold poultice for strains, sprains, sore shins, splints, bruises, capped hocks/elbows- Use as a dry dressing over open wounds or for pressure padding- Pre-cut hoof shape