Learn More
Med Vet International Animax Ointment (Panalog), 7.5mL
Catalog No. 502828808
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
7.5mL Animax ointment is a brand name for Nystatin, Ceomycin Sulfate, Thiostrpton, and Triamcinolone Acetonide. This product is used with dogs and cats and is classified as an antibiotic.
Catalog No. 50-282-8808 Supplier Med Vet International Supplier No. RXANIM7.5ML
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
7.5mL Animax ointment is a brand name for Nystatin, Ceomycin Sulfate, Thiostrpton, and Triamcinolone Acetonide. This product is used with dogs and cats and is classified as an antibiotic.