Learn More
Inorganic Ventures 7 ANION CALIBRATION STD 125mL
SDP
Catalog No. 50723581
Encompass_Preferred
Matrix: Water (H2O), Instrument(s): Common Multi-Ion Standards, Analyte: NO3-, NO2-, and Br- 100 ug/mL each, Cl- 30 ug/mL, F- 20 ug/mL, SO4-2, and PO4-3 150 ug/mL each.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search
Catalog No. 50-723-581 Supplier Inorganic Ventures Supplier No. ICFAS1A125ML
Matrix: Water (H2O), Instrument(s): Common Multi-Ion Standards, Analyte: NO3-, NO2-, and Br- 100 ug/mL each, Cl- 30 ug/mL, F- 20 ug/mL, SO4-2, and PO4-3 150 ug/mL each.
ATTENTION: Inorganic Ventures is no longer supplying paper copies of Certificate of Analyses(CoA) and Safety Data Sheets (SDS). Please use this link to download your copies: https://www.inorganicventures.com/coa-sds-search