Learn More
Epoch Life Science Inc GenCatch™ Anion Exchange DNA Midi Columns (50/PK)
SDP
Catalog No. NC1214288
Encompass_Preferred
Ion exchange columns for transfection grade plasmid DNA purification. Anion-exchange technology. 100 ug plasmid DNA binding capacity. Gravity flow. Compatible with third party buffer sets.
Catalog No. NC1214288 Supplier Epoch Life Science Inc Supplier No. 1970050
Ion exchange columns for transfection grade plasmid DNA purification. Anion-exchange technology. 100 ug plasmid DNA binding capacity. Gravity flow. Compatible with third party buffer sets.