Learn More
Crescent Chemical Co Inc ANION STANDARD MIX 1 100ML
SDP
Catalog No. 5072379
Encompass_Preferred
Anion Standard Mix 1, 6 Components In Water Multi Ion Chromatography Standard - Bromide, Chloride, Fluoride, Nitrate, Phosphate, Sulfate 100 ug/mL 100mL
Catalog No. 50-723-79 Supplier Crescent Chemical Co Inc Supplier No. IMAIM001
Anion Standard Mix 1, 6 Components In Water Multi Ion Chromatography Standard - Bromide, Chloride, Fluoride, Nitrate, Phosphate, Sulfate 100 ug/mL 100mL