Learn More
Epoch Life Science Inc GenCatch™ Anion Exchange DNA Maxi Columns (10/PK)
SDP
Catalog No. 501538103
Encompass_Preferred
Ion exchange columns for transfection grade plasmid DNA purification. Anion-exchange technology. 1000 ug (1 mg) plasmid DNA binding capacity. Gravity flow. Compatible with third party buffer sets.
Catalog No. 50-153-8103 Supplier Epoch Life Science Inc Supplier No. 1980010
Ion exchange columns for transfection grade plasmid DNA purification. Anion-exchange technology. 1000 ug (1 mg) plasmid DNA binding capacity. Gravity flow. Compatible with third party buffer sets.