Learn More
Waters Corp Waters Anionic Polar Pesticide Column, 130Å, 5 μm, 2.1 mm x 150 mm
Supplier:  Waters Corp 186009288
Encompass_Preferred
The Waters Anionic Polar Pesticide Column is specifically designed for quick and reproducible testing of highly anionic polar pesticides in agricultural products and finished foods. It addresses the analytical challenges posed by zwitterionic and highly polar pesticides like Glyphosate by providing application-specific column chemistry for ease of use, faster results, and reliable low-level detection, all while maintaining key analyte separations.
  • Extended peak shape integrity
  • Maintain key analyte separations
  • Retention without derivatization
  • Reliable low-level detection – no need for non-MS friendly buffers
Catalog No. 50-203-8973